Learn More
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
£216.08 - £759.00
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
IUPAC Name | boranylidyneamine |
SMILES | B=N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15432377
|
Thermo Scientific Alfa Aesar
045573.KG |
1 Pc. |
£216.08
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15442377
|
Thermo Scientific Alfa Aesar
045573.KJ |
5 Pc. | N/A | ||||||
Description
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
boranylidyneamine |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 | |
B=N |
Specifications
10043-11-5 | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
boranylidyneamine | |
66227 | |
24.82 | |
Bag |
Hot Pressed Rod | |
MFCD00011317 | |
Insoluble in water. Boron nitride is insoluble in the acids. Soluble in alkaline molten salts and nitrides, | |
B=N | |
24.82 | |
CHEBI:50883 | |
99.5% | |
Boron nitride |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
missing translation for 'einecsNumber' : 233-136-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only