Learn More
Boron nitride paste, Thermo Scientific Chemicals
Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength
£43.62 - £817.00
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
SMILES | B=N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11350408
|
Thermo Scientific Alfa Aesar
043773.14 |
25 g |
£43.62
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360408
|
Thermo Scientific Alfa Aesar
043773.22 |
100 g |
£127.64
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340408
|
Thermo Scientific Alfa Aesar
043773.36 |
500 g |
£507.53
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370408
|
Thermo Scientific Alfa Aesar
043773.A3 |
2 kg | N/A | ||||||
Description
Resistance against molten metals and salts, high thermal conductivity and dielectric strength
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite rShow More | |
CHEBI:50883 |
Specifications
10043-11-5 | |
Paste | |
MFCD00011317 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
azanylidyneborane | |
66227 | |
Odorless | |
1.4 |
White | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
B=N | |
24.82 | |
CHEBI:50883 | |
White paste | |
Boron nitride |
Safety and Handling
H319-H335: Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.