Learn More
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals
£31.80 - £241.00
Chemical Identifiers
CAS | 1303-86-2 |
---|---|
Molecular Formula | B2O3 |
Molecular Weight (g/mol) | 69.62 |
MDL Number | MFCD00011315 |
InChI Key | JKWMSGQKBLHBQQ-UHFFFAOYSA-N |
Synonym | boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g |
PubChem CID | 518682 |
ChEBI | CHEBI:30163 |
SMILES | O=BOB=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391639
|
Thermo Scientific Alfa Aesar
012290.36 |
500 g |
£31.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311649
|
Thermo Scientific Alfa Aesar
012290.A3 |
2 kg |
£71.90
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301649
|
Thermo Scientific Alfa Aesar
012290.A9 |
10 kg |
£241.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1303-86-2 | |
69.62 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
518682 | |
O=BOB=O |
B2O3 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
CHEBI:30163 |
Specifications
1860°C | |
1303-86-2 | |
B2O3 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo[(oxoboranyl)oxy]borane | |
518682 | |
69.62 | |
2.46 ^+ 0.01 g/mL |
∽450°C | |
97.5% min., typically 98.5% | |
MFCD00011315 | |
Slightly soluble in water and methanol. | |
O=BOB=O | |
69.62 | |
CHEBI:30163 | |
Hygroscopic | |
Boron oxide |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H360FD
missing translation for 'einecsNumber' : 215-125-8
missing translation for 'rtecsNumber' : ED7900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only