missing translation for 'onlineSavingsMsg'
Learn More
Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals Product Code.: 10628894

Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals

Product Code. 10628894
5 kg, Plastic Drum
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10628894

Brand: Thermo Scientific Acros 206300050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500gShow MoreShow Less
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo[(oxoboranyl)oxy]borane
SMILES O=BOB=O

Specifications

Boiling Point 1860.0°C
Melting Point 450.0°C
Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98%
Linear Formula B2O3
Fieser 01,673; 02,32; 03,33; 09,1349
Merck Index 15,1348
Solubility Information Solubility in water: 36g/L (25°C). Other solubilities: soluble in hot water,soluble in alcohol,glycerol and acids
Formula Weight 69.61
Percent Purity 98%
Packaging Plastic Drum
Specific Gravity 2.46
Density 2.4600g/mL
Chemical Name or Material Boron oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466861 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number206300050,206300010
Lot NumberA0461815 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number206300050,206300010
Lot NumberA0459961 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog Number206300050,206300250,206300010
Lot NumberA0456988 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number206300050,206300250,206300010
Lot NumberA0455944 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog Number206300050,206300250,206300010
compliance-icons
Product Identifier
  • Boron oxide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.