Learn More
Boron pieces, crystalline, 1-30mm (0.039-1.2in), 99.4% (metals basis), Thermo Scientific Chemicals
£120.60 - £1761.77
Chemical Identifiers
CAS | 7440-42-8 |
---|---|
Molecular Formula | B |
Molecular Weight (g/mol) | 10.81 |
MDL Number | MFCD00134034 MFCD00151272 |
InChI Key | ZOXJGFHDIHLPTG-UHFFFAOYSA-N |
Synonym | boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyneShow More |
PubChem CID | 5462311 |
ChEBI | CHEBI:27560 |
IUPAC Name | boron |
SMILES | [B] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354827
|
Thermo Scientific Alfa Aesar
041858.09 |
10 g |
£120.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364827
|
Thermo Scientific Alfa Aesar
041858.18 |
50 g |
£496.47
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344827
|
Thermo Scientific Alfa Aesar
041858.30 |
250 g |
£1761.77
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-42-8 | |
10.81 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
5462311 | |
boron |
B | |
MFCD00134034 MFCD00151272 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyneShow More | |
CHEBI:27560 | |
[B] |
Specifications
7440-42-8 | |
Crystalline Pieces | |
B | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
[B] | |
10.81 | |
CHEBI:27560 | |
Odorless | |
Boron |
Gray | |
99.4% (Metals basis) | |
MFCD00134034 MFCD00151272 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
boron | |
5462311 | |
700°C (1292°F) | |
Dark Gray pieces |
Safety and Handling
H302; Harmful if swallowed.
missing translation for 'einecsNumber' : 231-151-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.