missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

£101.51 - £534.66

Specifications

Chemical Name or Material Boron tribromide
CAS 10294-33-4, 75-09-2
Molecular Formula BBr3
Linear Formula BBr3
MDL Number MFCD00011312
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10525334
View Documents
Thermo Scientific Acros
427101000
100 mL AcroSeal™ Glass bottle
£101.51
100mL
Please to purchase this item. Need a web account? Register with us today!
10336774
View Documents Promotion Details
Thermo Scientific Acros
427108000
800 mL AcroSeal™ Glass bottle
£534.66
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Boron tribromide
BBr3
MFCD00011312
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
ILAHWRKJUDSMFH-UHFFFAOYSA-N
tribromoborane
25134
AcroSeal™ Glass bottle
1.4670g/mL
10294-33-4, 75-09-2
BBr3
15,1349
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4
BrB(Br)Br
250.52
250.52
1.467
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Suspected of causing cancer.
Reacts violently with water.
May cause drowsiness or dizziness.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-657-9

Product Identifier
  • Boron tribromide, 1M solution in methylene chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H300-Fatal if swallowed.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only