Learn More
Boron tribromide, 99.9%, Thermo Scientific Chemicals
CAS: 10294-33-4 | BBr3 | 250.52 g/mol
£49.95 - £440.19
Chemical Identifiers
CAS | 10294-33-4 |
---|---|
Molecular Formula | BBr3 |
Molecular Weight (g/mol) | 250.52 |
MDL Number | MFCD00011312 |
InChI Key | ILAHWRKJUDSMFH-UHFFFAOYSA-N |
Synonym | boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide |
PubChem CID | 25134 |
IUPAC Name | tribromoborane |
SMILES | BrB(Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10389400
|
Thermo Scientific
295200250 |
25 g | Glass bottle |
£49.95
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10481491
|
Thermo Scientific Acros
295201000 |
100 g | Glass bottle |
£130.65
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10237603
|
Thermo Scientific Acros
295205000 |
500 g | Glass bottle |
£440.19
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acidChemical Identifiers
10294-33-4 | |
250.52 | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
25134 | |
BrB(Br)Br |
BBr3 | |
MFCD00011312 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
tribromoborane |
Specifications
91.2°C | |
10294-33-4 | |
Liquid | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol | |
BrB(Br)Br | |
250.52 | |
250.52 | |
2.65 | |
Boron tribromide |
-46.0°C | |
Amber | |
99.85% min. (By difference) | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
99.9% | |
2.6500g/mL |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-657-9
- Boron tribromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.