missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 99+%, Thermo Scientific Chemicals

Product Code. 10281852 Shop All Thermo Scientific Products
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-33-4
Molecular Formula BBr3
Molecular Weight (g/mol) 250.52
MDL Number MFCD00011312
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromideShow MoreShow Less
PubChem CID 25134
IUPAC Name tribromoborane
SMILES BrB(Br)Br

Specifications

Boiling Point 91.2°C
Melting Point -46.0°C
Quantity 100 g
Assay Percent Range 99+%
Linear Formula BBr3
Fieser 01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43
Merck Index 15, 1349
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol
Formula Weight 250.52
Percent Purity ≥99%
Packaging Glass bottle
Specific Gravity 2.65
Density 2.6500g/mL
Chemical Name or Material Boron tribromide, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459144 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number365215000,365211000,365210250
Lot NumberA0468378 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number365215000,365211000,365210250
Lot NumberA0459144 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number365215000,365211000,365210250
Lot NumberA0448139 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number365215000,365211000,365210250
Lot NumberA0428313 Certificate TypeCertificate of Analysis Date01/11/2023 Catalog Number365215000,365211000,365210250
compliance-icons compliance-icons
Product Identifier
  • Boron tribromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron tribromide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.