Learn More
Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 428328000
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron tribromide dimethyl sulfide | |
29957-59-3, 75-09-2 | |
C2H6BBr3S | |
tribromoborane-methyl sulfide, boron tribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex, tribromo dimethyl-??-sulfanylidene-??-borane, dimethyl sulfide-tribromoborane, tribromo dimethylsulfonio boranuide, boron tribromide-methylsulfide complex, borontribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex solution | |
YNLFEVAOQLXINF-UHFFFAOYSA-N | |
(methylsulfanyl)methane; tribromoborane | |
4181510 | |
AcroSeal™ Glass Bottle | |
1.4560g/mL |
1M solution in methylene chloride | |
800 mL | |
MFCD00043296 | |
Solubility in water: reacts | |
CSC.BrB(Br)Br | |
312.65 | |
312.66 | |
1.456 |
Safety and Handling
- Boron tribromide dimethyl sulfide, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only