missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 176688000


 View more versions of this product

Product Code. 10332963

  • £282.41 / 800mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Boron trichloride
1M Solution in Methylene Chloride
BCl3
MFCD00011313
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44
15, 1350
Solubility in water: may decompose.
ClB(Cl)Cl
117.16
117.17
1.326
Liquid
Colorless to Brown
10294-34-5, 75-09-2
BCl3
0.9 to 1.1M
1.3260g/mL
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
FAQYAMRNWDIXMY-UHFFFAOYSA-N
trichloroborane
25135
AcroSeal™ Glass Bottle
800 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Boron trichloride, 1M solution in methylene chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H301-Toxic if swallowed.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only