missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron Trichloride, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Boron trichloride | 10294-34-5 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Physical Form | Powder/Solid |
CAS | 10294-34-5, 75-09-2 |
CAS Min % | 0.1, 99.9 |
CAS Max % | 0.1, 99.9 |
Molecular Formula | BCl3 |
MDL Number | MFCD00011313 |
InChI Key | FAQYAMRNWDIXMY-UHFFFAOYSA-N |
SMILES | ClB(Cl)Cl |
IUPAC Name | trichloroborane |
Molecular Weight (g/mol) | 117.16 |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'