missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride, 12% (1.5M) in methanol

Product Code. 10186840 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 10186840

Brand: Thermo Scientific Acros 402765000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Boron trifluoride, 12% (1.5M) in methanol, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3) | CAS: 373-57-9 | BF3 | 67.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent, Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
Name Note 1.5M solution in methanol
CAS 373-57-9, 67-56-1
Color Amber to Colorless
Physical Form Solution
Quantity 500 g
Molecular Formula BF3
Linear Formula BF3
MDL Number MFCD00011316
Flash Point 4°C
Merck Index 14, 1349
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
Solubility Information Solubility in water: may decompose
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILES FB(F)F
IUPAC Name trifluoroborane
Molecular Weight (g/mol) 67.81
PubChem CID 11062313
Formula Weight 67.81
Percent Purity 10 to 15% (w/v BF3)
Packaging Glass bottle
Specific Gravity 0.87
Density 0.8700g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride, 12% in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.