missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
Melting Point | -20.0°C |
Color | Colorless to Yellow |
Density | 1.2220g/mL |
Boiling Point | 59.0°C (4.0 mmHg) |
Flash Point | 68°C |
Packaging | Glass bottle |
Linear Formula | BF3·2CH3OH |
Refractive Index | 1.34 |
Quantity | 1 kg |
Specific Gravity | 1.222 |
Show More |
Safety and Handling
Product Identifier
- Boron trifluoride dimethanol complex
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Methyl alcohol, Borate(1-), trifluoromethoxy-, (T-4)-, hydrogen, compound with methanol (1:1), Boron
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'