missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11396169
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-43-5
Molecular Formula C2H6BF3S
Molecular Weight (g/mol) 129.94
MDL Number MFCD00013193
InChI Key BRWZPVRDOUWXKE-UHFFFAOYSA-N
Synonym boron fluoride-dimethyl sulfide complex, dimethyl-??-sulfanylidene trifluoro-??-borane, trifluoroborane-methyl sulfide, dimethyl sulfide-trifluoroborane, trifluoro methylsulfanyl methane boron, boron trifluoride methyl sulfide complex, boron trifluoride-methyl sulfide complex, boron trifluoride-dimethyl sulfide complex, purified, packaged under argon in resealable chemseal bottles
PubChem CID 71311438
IUPAC Name dimethylsulfonio(trifluoro)boranuide
SMILES CSC.FB(F)F

Specifications

Physical Form Liquid
Quantity 500 mL
UN Number UN2920
Formula Weight 129.94
Packaging ChemSeal™ bottles
Sensitivity Moisture Sensitive
Density 1.235g/mL
Chemical Name or Material Boron trifluoride-dimethyl sulfide complex, Purified

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'