missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride etherate, approx. 48% BF3, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Melting Point | -60.0°C |
Color | Colorless to Straw-Yellow |
Density | 1.1200g/mL |
Boiling Point | 126.0°C |
Flash Point | 58°C |
Packaging | Glass bottle |
Linear Formula | (C2H5)2O·BF3 |
Refractive Index | 1.3429 to 1.3449 |
Quantity | 100 g |
Fieser | 01,70; 02,35; 03,33; 04,44; 05,52; 06,65; 07,31; 08,51; 09,64; 10,52; 11,72; 12,66; 13,3; 14,54; 15,45; 16,44; 17,52 |
Show More |
Safety and Handling




Product Identifier
- Boron trifluoride etherate
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boron trifluoride etherate, approx. 48% BF3, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction