Learn More
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™, Thermo Scientific Chemicals
£25.70 - £68.50
Chemical Identifiers
CAS | 109-63-7 |
---|---|
Molecular Formula | C4H10BF3O |
Molecular Weight (g/mol) | 141.93 |
MDL Number | MFCD00013194 |
InChI Key | KZMGYPLQYOPHEL-UHFFFAOYSA-N |
Synonym | Boron trifluoride ethyl ether |
PubChem CID | 8000 |
IUPAC Name | ethoxyethane;trifluoroborane |
SMILES | FB(F)F.CCOCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10304034
|
Thermo Scientific Acros
427111000 |
100 mL | AcroSeal™ Glass Bottle |
£25.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396814
|
Thermo Scientific Acros
427118000 |
800 mL | AcroSeal™ Glass Bottle |
£68.50
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
109-63-7 | |
141.93 | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
8000 | |
FB(F)F.CCOCC |
C4H10BF3O | |
MFCD00013194 | |
Boron trifluoride ethyl ether | |
ethoxyethane;trifluoroborane |
Specifications
109-63-7 | |
Colorless to Straw-Yellow | |
126.0°C | |
C4H10BF3O | |
1.3429 to 1.3449 | |
15, 1352 | |
Boron trifluoride ethyl ether | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
ethoxyethane;trifluoroborane | |
8000 | |
141.93 | |
Liquid |
-60.0°C | |
1.1200g/mL | |
58°C | |
(C2H5)2O·BF3 | |
MFCD00013194 | |
1.12 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether | |
FB(F)F.CCOCC | |
141.93 | |
1.89 mPa.s (20°C) | |
ca. 48% BF3 | |
Boron trifluoride etherate, ca. 48% BF3 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-689-8
- Boron trifluoride etherate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only