missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-tetrahydrofuran complex, BF{3} 45.5%, Thermo Scientific Chemicals

Product Code. 11348536
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Boron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.

Solubility
Miscible with dimethyl formamide.

Notes
Incompatible with strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-34-0
Molecular Formula C4H8BF3O
Molecular Weight (g/mol) 139.91
MDL Number MFCD00040372
InChI Key CHNLPLHJUPMEOI-UHFFFAOYSA-N
Synonym boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor
PubChem CID 60146129
SMILES FB(F)F.C1CCOC1

Specifications

Melting Point 8°C to 9°C
Density 1.268
Boiling Point 180°C
Flash Point 92°C (197°F)
Quantity 500 g
UN Number UN2922
Sensitivity Moisture sensitive
Solubility Information Miscible with dimethyl formamide.
IUPAC Name trifluoro(oxolan-1-ium-1-yl)boranuide
Formula Weight 139.91
Assay BF3 45.5%
Chemical Name or Material Boron trifluoride-tetrahydrofuran complex
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'