Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Shop All Brand Products
Click to view available options
Size:
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 45/40
NS 7/16
Unit Size:
Each
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Hollow
- Hexagonal grip
Specifications
Specifications
| Quantity | 1/Pk. |
| Size | NS 45/40 |
| Standard Taper Joint | NS 45/40 |
| Material | Borosilicate Glass |
| Product Type | Ground Joint Stopper |
| Shape | Conical |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion