missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass 3.3 Lid with Socket

Borosilicate Glass 3.3 Lid with Socket Lid designed for use with desiccators.

Brand:  BRAND™ 65743

171.70 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10059910

  • £192.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes a socket in the lid
  • Nomical size: 250 mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

320 mm
Lid
Borosilicate Glass (3.3)
NS 24/29 socket
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers