missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Borosilicate Glass Class A Mortar
Brand: Wheaton™ 358028
133.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Autoclavable | |
2 mL | |
Mortar | |
45mm |
11mm | |
Borosilicate Glass | |
For use in preparation of tissue homogenates and other similar applications |