missing translation for 'onlineSavingsMsg'
Learn More

Conical Filter Vacuum Flask with Hose Tubulature

Use this Conical Filter Vacuum Flask with Hose Tubulature with a vacuum funnel and filter. This thick-walled Ehlenmeyer flask features a glass tube protrusion about an inch from its neck and is also known as a Büchner flask.

Brand:  N/A FHD-345-110J

Code : 01

Additional Details : Weight : 1.05750kg

 View more versions of this product

Product Code. 12373886

  • £152.74 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • With 8mm outer diameter tubulature
  • Thick-walled borosilicate glass
  • Vacuum flask
Specifications

Specifications

45 mm
8 mm
2000 mL
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers