Learn More
BRAND™ Borosilicate Glass Conical Ground Joint Stopper
Solid or semi-hollow with octagonal grip. BRAND™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3. to DIN 12252 standards.
£10.35 - £22.75
Specifications
Material Borosilicate Glass
Product Type Ground Joint Stopper
Shape Conical
View More Specs
Products 5
Product Code Brand Description Standard Taper Joint Price Quantity & Availability  
Product Code Brand Description Standard Taper Joint Price Quantity & Availability  
10273821
View Documents
BRAND™
144520
Solid with octagonal grip NS 14/23
£10.85
Each
In Stock
to see stock.
Add to basket
10798621
View Documents
BRAND™
144525
Solid with octagonal grip NS 19/26
£11.55
Each
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
10507461
View Documents
BRAND™
144535
Semi-hollow with octagonal grip NS 29/32
£22.10
Each
In Stock
to see stock.
Add to basket
10243681
View Documents
BRAND™
144515
Solid with octagonal grip NS 12/21
£10.35
Each
In Stock
to see stock.
Add to basket
10643012
View Documents
BRAND™
144540
Semi-hollow with octagonal grip NS 34/35
£22.75
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
  • Solid or semi-hollow
  • Octagonal grip
Specifications
Specifications
Borosilicate Glass
Ground Joint Stopper
Conical