Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
£73.53 - £188.24
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10643161
|
Lenz Laborglasinstrumente™
04006137 |
100 mL |
£82.99
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11852672
|
Lenz Laborglasinstrumente™
04006170 |
1000 mL |
£161.20
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||||
11836084
|
Lenz Laborglasinstrumente™
04006173 |
2000 mL |
£188.24
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
10100121
|
Lenz Laborglasinstrumente™
04006149 |
250 mL |
£120.55
Pack of 2 |
In Stock
Log in to see stock. |
||||||
10393511
|
Lenz Laborglasinstrumente™
04006128 |
50 mL |
£73.53
Pack of 2 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11871593
|
Lenz Laborglasinstrumente™
04006158 |
500 mL |
£117.52
Pack of 2 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
- With scale (approx. volume/mL)
- With PTFE plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Polyethylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale