Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04000073
This item is not returnable.
View return policy
Description
- Without scale; with solid glass plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
2000 mL | |
Borosilicate Glass | |
Polyethylene |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale > Capacity: 2000mL
Spot an opportunity for improvement?Share a Content Correction