missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Filter Flask

Lateral socket for vacuum tubing with outer diameter 15-18 mm

Brand:  BRAND™ 73719

49.22 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 0.47000kg

 View more versions of this product

Product Code. 10325331

  • £56.25 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

  • Boro 3.3 glass
  • DIN 12476, ISO 6556
  • Specifications

    Specifications

    Clear
    500 mL
    Borosilicate Glass
    Lateral socket
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers