missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Borosilicate Glass Filter Flask
Lateral socket for vacuum tubing with outer diameter 15-18 mm
Brand: BRAND™ 73724
60.13 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.81000kg
Description
Specifications
Clear | |
1000 mL | |
Lateral socket |
18 mm | |
Borosilicate Glass |