missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Filter Flask

Lateral socket for vacuum tubing with outer diameter 15-18 mm

Brand:  BRAND™ 73729

159.45 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.



Additional Details : Weight : 1.34000kg

 View more versions of this product

Product Code. 10759201

  • £170.00 / Each
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

  • Boro 3.3 glass
  • DIN 12476, ISO 6556
  • Specifications

    Specifications

    DIN 12 476, ISO 6556
    60mm
    Borosilicate Glass
    Lateral socket
    1/Pk.
    Clear
    2000 mL
    265 mm
    Buchner filter flask
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers