Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
£27.00 - £84.00
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10120181
|
Lenz Laborglasinstrumente™
01400045 |
NS 45/40 |
£84.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
10517371
|
Lenz Laborglasinstrumente™
01400034 |
NS 34/35 |
£49.54
Pack of 5 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
10291631
|
Lenz Laborglasinstrumente™
01400007 |
NS 7/16 |
£27.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12252, form C
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Hollow Stopper |