missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPE Scientific™ Borosilicate Glass J Young NMR Valves
Brand: GPE Scientific™ S-5-600-JY-7-YELLOW
Code : 50
Additional Details : Weight : 1.00000kg
Description
Order Info
LengthMetric
Specifications
J Young NMR Valve | |
178 mm | |
Female joint |
Borosilicate Glass | |
600MHz tube | |
1 Each |