Learn More
Quickfit™ Borosilicate Glass Markham Distillation Assembly
Quickfit™ Borosilicate Glass Markham Distillation Assembly is used for volatile fatty acid tests and can be used to carry out a large number of distillations in rapid successioin
Brand: Quickfit™ 46MC
457.22 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Complies with BS1428: Part B2
- Enables a large number of distillations to be carried out in rapid succession
- Comprises Markham still body, Liebig condenser and connecting joint clip
- Still has evaporator vessel enclosed in a steam jacket
- When connected to a suitable steam supply the steam passes through the sample to promote distillation
- When steam supply is shut off the apparatus automatically sucks back any residue in the vessel thus preparing itself for the next determination
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Enables a large number of distillations to be carried out in rapid succession | |
Markham distillation assembly | |
Borosilicate Glass | |
BS 1428: Part B2 |
Product Suggestions
Customers who viewed this item also viewed