missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Borosilicate Glass Medium Wall Rimless Test Tube

Manufactured from borosilicate glass with a medium wall design. Fisherbrand™ Borosilicate Glass Medium Wall Rimless Test Tubes are ideal for chemical and biological work.

£28.78 - £58.19

Specifications

Certifications/Compliance ISO 4142
Material Borosilicate Glass
Quantity 100 Pack
Rimmed No
View More Specs

Products 6
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
11912208
View Documents Promotion Details
Fisherbrand™
FB63141
10 mm 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11922208
View Documents Promotion Details
Fisherbrand™
FB63142
12 mm 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11942208
View Documents Promotion Details
Fisherbrand™
FB63144
16 mm 100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11952208
View Documents Promotion Details
Fisherbrand™
FB63145
16 mm 125 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11972208
View Documents Promotion Details
Fisherbrand™
FB63147
18 mm 150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12088099
View Documents Promotion Details
Fisherbrand™
FB63146
16 mm 150 mm
£49.67
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Features: rimless medium walls
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
Specifications

Specifications

ISO 4142
Borosilicate Glass
100 Pack
No
Videos
SDS
Documents

Documents

Product Certifications