Learn More
Fisherbrand™ Glass Square Coverslips
Glass Square Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
Brand: Fisherbrand™ MCS02222
Description
- Manufactured from clear soda glass
- Consistent in cut, thickness and dimensional accuracy
- Free from blisters and flaws
Specifications
Coverslip | |
Soda Lime Glass | |
22 mm | |
200/Pk. |
Square | |
22 mm | |
#1.5 (0.16 to 0.19 mm) |