missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime Glass Petri Dishes

Product Code. 16190741 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
60 mm
80 mm
100 mm
120 mm
150 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Each
Pack of 18
Pack of 20
This item is not returnable. View return policy

Product Code. 16190741

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-quality petri dish, with lid.

  • Constructed of soda lime glass.
  • Includes lid

Specifications

Closure Included Closure Included
Closure Material Soda Lime Glass
Material Soda Lime Glass
Shape Round
Type Petri Dish
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.