Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01400024
 View more versions of this product
Product Code. 10550052
  • £24.25 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow
Thread Size: 24/29 Cone NS
Hollow Stopper
10/Pk.
Borosilicate Glass