missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Potter-Elvehjem Homogenizers

Designed specifically to work with softer tissue. Fisherbrand™ Potter-Elvehjem Homogenizers are manufactured from durable borosilicate glass.

Brand:  Fisherbrand™ 22008F

 View more versions of this product

Product Code. 15301331

  • £108.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Fully autoclavable
  • Available in a range of capacities
  • Grinding Clearance Pestle and Mortar of 0.076 - 0.126mm
  • Manufactured using precision bore glass with a PTFE pestle
  • Supplied with a stainless steel rod for use with an electric motor
  • Optional stainless steel handle for manual grinding is available separately (Mfr. No. 20310)
Specifications

Specifications

0.076 - 0.126mm
15 mL
Homogenizers
250mm
Autoclavable
Borosilicate Glass
80mm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.