Learn More
BRAND™ Borosilicate Glass Powder Funnel
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
Brand: BRAND™ 146516
10.55 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from borosilicate glass.
- Ground and fire-polished upper rim.
- Short, wide stem optimal for funneling powder and other ground materials.
Specifications
80 mm | |
22 mm | |
Borosilicate Glass |