Learn More
Day Impex™ Borosilicate Glass Refill
Designed with practicality and safety in mind. Day Impex™ Borosilicate Glass Refill is ultra-low evacuated to allow for safe and efficient handling of all liquified gases, freezing mixtures, and specimens.
£146.00 - £620.00
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
11877442
|
Day Impex™
210015 |
380 mL | 100 mm |
£146.00
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||||
11726409
|
Day Impex™
210002 |
2 L | 106 mm |
£157.00
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
11807452
|
Day Impex™
210017 |
1 L | 138 mm |
£219.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11817442
|
Day Impex™
210013 |
7 L | 200 mm |
£620.00
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
11736409
|
Day Impex™
210011 |
4500 mL | 150 mm |
£251.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11716409
|
Day Impex™
210001 |
1 L | 85 mm |
£217.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11837462
|
Day Impex™
210007 |
1500 mL | 90 mm |
£164.00
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
Description
- Borosilicate glass inner vessels
- Ultra-low evacuated
- Safe and efficient handling of all liquified gases, freezing-mixtures and specimens
Specifications
Borosilicate Glass |