Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04030049
 View more versions of this product
Product Code. 10213961
  • £192.00 / Pack of 2
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cylindrical shape with straight discharge tube
  • With NS socket and PE stopper
  • With solid glass plug
  • With Screwthreade retaining device
  • With scale
Specifications
Specifications
250 mL
Borosilicate Glass
Polyethylene