Learn More
Fisherbrand™ Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
Brand:  Fisherbrand™ 00793104
 View more versions of this product
Product Code. 10720295
  • £48.25 / Pack of 10
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Soda Lime glass.
  • Short stem funnel
  • Less likely to clog when funneling thicker substances.
Specifications
Specifications
45mm
Soda Lime Glass
Ideal for filtering applications in the laboratory
7mm
90 mm
50mm