missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Borosilicate Glass Spinner Flask
Features an adjustable paddle blade impeller, that does not protrude through the cap, for greater turbulence
£790.00 - £1300.00
Product Code | Brand | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
10260232
|
Wheaton™
356927 |
3 L | Flask Only |
£790.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10498612
|
Wheaton™
356889 |
6 L | Spinner Flask |
£1300.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
Description
- Features an adjustable paddle blade impeller, that does not protrude through the cap, for greater turbulence.
- The bottom dimple in 125mL and larger flasks prevents crushing of cells.
- 500mL and larger units have 45mm sidearms to be used as convenient vents and ports.
- All flasks provide a headspace ratio of 1:1 or greater to provide maximum surface interface between culture and vessel atmosphere.
- Unit consists of glass flask, PTFE and glass impeller assembly, and PTFE and silicone lined cap.
- Entire unit may be autoclaved.
- Optional replacement stainless steel shaft assemblies are available for each vessel.
- Sizes 125mL to 1,000mL are graduated in 50mL increments. 3, 000mL to 8, 000mL are graduated to 500mL increments.
- 25mL and 50mL are ungraduated.
- For microcarrier and suspension cultures such as insect cells, hybridomas and adapted cell lines.
Specifications
Borosilicate Glass |