Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
£81.00 - £141.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10078420
|
Lenz Laborglasinstrumente™
04014170 |
1000 mL |
£141.00
Pack of 2 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
10424111
|
Lenz Laborglasinstrumente™
04014173 |
2000 mL |
£93.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10567541
|
Lenz Laborglasinstrumente™
04014137 |
100 mL |
£81.00
Pack of 2 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10633161
|
Lenz Laborglasinstrumente™
04014149 |
250 mL |
£96.00
Pack of 2 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10415431
|
Lenz Laborglasinstrumente™
04014158 |
500 mL |
£91.37
Pack of 2 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10567351
|
Lenz Laborglasinstrumente™
04014128 |
50 mL |
£86.00
Pack of 2 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
- Without scale
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Polyethylene |