Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04016173
 View more versions of this product
Product Code. 11846164
  • £134.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • with PTFE plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifications
Specifications
2000 mL
Borosilicate Glass
Polyethylene