Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug are used to separate two immiscible liquids of different densities
£59.20 - £125.90
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10159930
|
Lenz Laborglasinstrumente™
04010070 |
1000 mL |
£125.90
Pack of 2 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10271721
|
Lenz Laborglasinstrumente™
04010037 |
100 mL |
£69.00
Pack of 2 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
10231821
|
Lenz Laborglasinstrumente™
04010049 |
250 mL |
£83.59
Pack of 2 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
10149930
|
Lenz Laborglasinstrumente™
04010058 |
500 mL |
£97.00
Pack of 2 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10242151
|
Lenz Laborglasinstrumente™
04010073 |
2000 mL |
£84.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
12018538
|
Lenz Laborglasinstrumente™
04010028 |
50 mL |
£59.20
Pack of 2 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
- With NS socket and PE stopper
- without scale, with solid glass plug
- with screwthread retaining device
- material: borosilicate glass 3.3
Specifications
Borosilicate Glass | |
Polyethylene |