Learn More
GPE Scientific™ Borosilicate Glass Stirrer
For use with overhead stirrer motors
Brand:  GPE Scientific™ 30502
 View more versions of this product
Product Code. 12977066
  • £91.75 / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with overhead stirrer motors
  • Non-whip and autoclavable
  • The PTFE blades are easily removed or replaced
Specifications
Specifications
Overhead Stirrer Motors
B24
Uni-Form Precision Glass Stirrers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products