Learn More
Borosilicate Glass Stopcock Adapter with Cone
Control flow rate. Quickfit Jointed Ware™ QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand:  Quickfit MF11/3/SC
73.70 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11322045
  • £134.16 / Each
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Control flow rate. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
  • With GP Rotaflo™ PTFE stopcock: - No lubrication required - No contaminating greases - Fine flow control - Positive closure
  • Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stopcock Adapter
Borosilicate Glass
24/29
Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock