missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Borosilicate Glass Superloops

Use for the introduction of large sample volumes into a pressurised fluid LC system. Cytiva Lifescience™ Borosilicate Glass Superloops work in conjunction with 7-port valves.

Brand:  Cytiva 18-1113-82

Product Code. 11360112

  • £983.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Superloop can be used in conjunction with 7-port valves. The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection. They may also be used to inject pulses of a specific buffer solution.
  • Flexible: inject large or small sample volumes.
  • High reproducibility and recovery.
  • No gradient dilution with internal movable seal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
1 Each
580 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Borosilicate Glass Superloops > Length: 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.