Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Product Code. 15822285 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
75 mm
150 mm
Diameter (Metric):
12 mm
24 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 15822285
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
  • Borosilicate glass test tubes, grade 3.3
  • All ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 150 mm
Material Borosilicate Glass
Quantity 50 Pack
Diameter (Metric) 24 mm