missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Borosilicate Glass Test Tubes, Blue Graduation
Test tubes with accurate scale made from heat resistant DURAN™ glass.
£131.00 - £146.00
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
11792293
|
Hirschmann™
6420166 |
20 mL | with NS 14/23 Polyethylene Stopper |
£135.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
11782293
|
Hirschmann™
6420160 |
10 mL | with NS 12/21 Polyethylene Stopper |
£131.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
11830522
|
Hirschmann™
6420170 |
25 mL | with NS 14/23 Polyethylene Stopper |
£146.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
13118029
|
Hirschmann™
6420165 |
20 mL | NS 14/23 Polyethylene Stopper |
£136.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- With NS-ground
- Poly stopper
- Blue graduation
Specifications
10 Pack | |
Yes | |
Test Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title