Learn More
Karl Hecht™ Assistent™ Glass Weighing Boats
For easy transfer of liquids and powders in the laboratory. Karl Hecht™ Assistent™ Glass Weighing Boats are manufactured from durable DURAN™ borosilicate 3.3 glass.
Brand:  Karl Hecht™ 44248010
 View more versions of this product
Product Code. 10489621
  • £15.15 / Each
Explore more special offers
Description
Description
  • Available in different capacities
  • Clear glass
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
10 mL
Borosilicate Glass
105 mm
Weighing Funnel