Learn More
Cytiva Borosilicate/Polyetheretherketone/Perfluor Rubber/Nitrilebutadiene Rubber Superloops
Use for the introduction of large sample volumes into a pressurised fluid LC system. Cytiva Lifescience™ Borosilicate/Polyetheretherketone/Perfluor Rubber/Nitrilebutadiene Rubber Superloops work in conjunction with 7-port valves.
Brand: Cytiva 18-1113-81
Code : GX
Additional Details : Weight : 0.90000kg
Description
- Superloop can be used in conjunction with 7-port valves. The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection. They may also be used to inject pulses of a specific buffer solution.
- Flexible: inject large or small sample volumes.
- High reproducibility and recovery.
- No gradient dilution with internal movable seal.
Specifications
1.58 mm | |
Aqueous Solutions and Alcohols | |
4°C to 40°C | |
30 mm |
30 mm dia. x 130 mm L | |
40 bar | |
130 mm | |
10 mL |